Learn More
4-(Dimethylamino)pyridine tribromide, 97%, Thermo Scientific Chemicals
$74.99 - $273.10
Chemical Identifiers
CAS | 92976-81-3 |
---|---|
Molecular Formula | C7H10Br3N2- |
Molecular Weight (g/mol) | 361.883 |
MDL Number | MFCD00043439 |
InChI Key | TYDZHQMKSLAFKM-UHFFFAOYSA-N |
Synonym | 4-dimethylamino pyridine hydrobromide perbromide |
PubChem CID | 131863682 |
SMILES | CN(C)C1=CC=NC=C1.Br[Br-]Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5501006
|
Thermo Scientific Chemicals
H5501006 |
5 g |
N/A
|
|
|||||
AAH5501014
|
Thermo Scientific Chemicals
H5501014 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
92976-81-3 | |
361.883 | |
TYDZHQMKSLAFKM-UHFFFAOYSA-N | |
131863682 |
C7H10Br3N2- | |
MFCD00043439 | |
4-dimethylamino pyridine hydrobromide perbromide | |
CN(C)C1=CC=NC=C1.Br[Br-]Br |
Specifications
92976-81-3 | |
C7H10Br3N2- | |
5 g | |
4-dimethylamino pyridine hydrobromide perbromide | |
CN(C)C1=CC=NC=C1.Br[Br-]Br | |
131863682 | |
97% |
129°C to 132°C | |
MFCD00043439 | |
UN3261 | |
TYDZHQMKSLAFKM-UHFFFAOYSA-N | |
361.883 | |
362.89 | |
4-(Dimethylamino)pyridine tribromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only