missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)stilbene 99.0+%, TCI America™
Supplier: TCI America D02555G
Specifications
| 4-(Dimethylamino)stilbene | |
| 149°C | |
| MFCD00017151 | |
| XGHHHPDRXLIMPM-CMDGGOBGSA-N | |
| N,N-dimethyl-4-[(E)-2-phenylethenyl]aniline | |
| 640024 | |
| ≥99.0% (T) |
| 1145-73-9 | |
| C16H17N | |
| 5 g | |
| CN(C)C1=CC=C(C=C1)C=CC2=CC=CC=C2 | |
| 223.319 | |
| 223.32 | |
| Crystalline Powder |
Chemical Identifiers
| 1145-73-9 | |
| 223.319 | |
| XGHHHPDRXLIMPM-CMDGGOBGSA-N | |
| N,N-dimethyl-4-[(E)-2-phenylethenyl]aniline |
| C16H17N | |
| MFCD00017151 | |
| 640024 | |
| CN(C)C1=CC=C(C=C1)C=CC2=CC=CC=C2 |
Safety and Handling
RTECSNumber : WJ3850000
TSCA : No