Learn More
4-Dimethylaminoazobenzene-4'-sulfonyl chloride, 98+%, Thermo Scientific Chemicals
$166.34 - $166.34
Chemical Identifiers
CAS | 56512-49-3 |
---|---|
Molecular Formula | C14H14ClN3O2S |
Molecular Weight (g/mol) | 323.80 |
MDL Number | MFCD00007444 |
InChI Key | VTVWTPGLLAELLI-UHFFFAOYSA-N |
Synonym | dabsyl chloride, 4-dimethylaminoazobenzene-4'-sulfonyl chloride, 4-dimethylamino azobenzene-4'-sulfonyl chloride, p-dimethylaminoazobenzene-4-sulfonyl chloride, p-p-dimethylamino phenyl azo benzenesulfonyl chloride, benzenesulfonyl chloride, 4-4-dimethylamino phenyl azo, benzenesulfonyl chloride, p-p-dimethylamino phenyl azo, 4-n,n-dimethylaminoazobenzene-4'-sulfonyl chloride, p-4-dimethylamino phenyl azo benzenesulphonyl chloride |
PubChem CID | 91660 |
IUPAC Name | 4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonyl chloride |
SMILES | CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL06057ME
|
Thermo Scientific Chemicals
L06057ME |
500 mg |
Each for $166.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56512-49-3 | |
323.80 | |
VTVWTPGLLAELLI-UHFFFAOYSA-N | |
91660 | |
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O |
C14H14ClN3O2S | |
MFCD00007444 | |
dabsyl chloride, 4-dimethylaminoazobenzene-4'-sulfonyl chloride, 4-dimethylamino azobenzene-4'-sulfonyl chloride, p-dimethylaminoazobenzene-4-sulfonyl chloride, p-p-dimethylamino phenyl azo benzenesulfonyl chloride, benzenesulfonyl chloride, 4-4-dimethylamino phenyl azo, benzenesulfonyl chloride, p-p-dimethylamino phenyl azo, 4-n,n-dimethylaminoazobenzene-4'-sulfonyl chloride, p-4-dimethylamino phenyl azo benzenesulphonyl chloride | |
4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonyl chloride |
Specifications
56512-49-3 | |
C14H14ClN3O2S | |
500 mg | |
3064095 | |
dabsyl chloride, 4-dimethylaminoazobenzene-4'-sulfonyl chloride, 4-dimethylamino azobenzene-4'-sulfonyl chloride, p-dimethylaminoazobenzene-4-sulfonyl chloride, p-p-dimethylamino phenyl azo benzenesulfonyl chloride, benzenesulfonyl chloride, 4-4-dimethylamino phenyl azo, benzenesulfonyl chloride, p-p-dimethylamino phenyl azo, 4-n,n-dimethylaminoazobenzene-4'-sulfonyl chloride, p-4-dimethylamino phenyl azo benzenesulphonyl chloride | |
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O | |
323.80 | |
323.8 | |
4-Dimethylaminoazobenzene-4'-sulfonyl chloride |
∽185°C (decomposition) | |
MFCD00007444 | |
UN3261 | |
Moisture sensitive | |
VTVWTPGLLAELLI-UHFFFAOYSA-N | |
4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonyl chloride | |
91660 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 260-235-1
RTECSNumber : DB8927800
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only