missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A117120E

 View more versions of this product

Catalog No. AAA117120E


Explore available promotions

Description

Description

4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

Solubility
Soluble in alcohol, ether, chloroform, acetic acid. Insoluble in water.

Notes
Light sensitive. Stable at room temperature.
Specifications

Specifications

4-Dimethylaminobenzaldehyde
73°C to 75°C
164°C (327°F)
C9H11NO
2500 g
Air and light sensitive
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-(dimethylamino)benzaldehyde
7479
98%
100-10-7
176°C to 177°C (17 mmHg)
Strong
MFCD00003381
606802
14,3230
Soluble in alcohol,ether,chloroform,acetic acid. Insoluble in water.
CN(C)C1=CC=C(C=O)C=C1
149.19
149.19

Chemical Identifiers

100-10-7
149.19
BGNGWHSBYQYVRX-UHFFFAOYSA-N
7479
CN(C)C1=CC=C(C=O)C=C1
C9H11NO
MFCD00003381
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
4-(dimethylamino)benzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 202-819-0

RTECSNumber : CU5775000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only