missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Dimethylaminobenzoic anhydride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3250303

 View more versions of this product

Catalog No. AAH3250303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Dimethylaminobenzoic anhydride
149°C to 154°C
MFCD09757554
4-dimethylaminobenzoic anhydride, 4-dimethylamino benzoyl 4-dimethylamino benzoate, acmc-20amjw, benzoic acid, 4-dimethylamino-, anhydride, 4-dimethylamino phenyl anhydride #, bis 4-dimethylamino benzoic anhydride, benzoic acid,4-dimethylamino-, anhydride with 4-dimethylamino benzoic acid
CN(C)C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)N(C)C
312.369
312.37
7474-31-9
C18H20N2O3
1 g
KGUATYWLOBCNMI-UHFFFAOYSA-N
[4-(dimethylamino)benzoyl] 4-(dimethylamino)benzoate
344732
97%

Chemical Identifiers

7474-31-9
312.369
KGUATYWLOBCNMI-UHFFFAOYSA-N
344732
CN(C)C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)N(C)C
C18H20N2O3
MFCD09757554
4-dimethylaminobenzoic anhydride, 4-dimethylamino benzoyl 4-dimethylamino benzoate, acmc-20amjw, benzoic acid, 4-dimethylamino-, anhydride, 4-dimethylamino phenyl anhydride #, bis 4-dimethylamino benzoic anhydride, benzoic acid,4-dimethylamino-, anhydride with 4-dimethylamino benzoic acid
[4-(dimethylamino)benzoyl] 4-(dimethylamino)benzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only