Learn More
4-(Diphenylamino)benzaldehyde, 98%, Thermo Scientific Chemicals
$52.37 - $755.42
Chemical Identifiers
CAS | 4181-05-9 |
---|---|
Molecular Formula | C19H15NO |
Molecular Weight (g/mol) | 273.34 |
MDL Number | MFCD00145131 |
InChI Key | UESSERYYFWCTBU-UHFFFAOYSA-N |
Synonym | 4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde |
PubChem CID | 77846 |
IUPAC Name | 4-(N-phenylanilino)benzaldehyde |
SMILES | C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6488703
|
Thermo Scientific Chemicals
H6488703 |
1 g |
Each for $52.37
|
|
|||||
AAH6488706
|
Thermo Scientific Chemicals
H6488706 |
5 g |
Each for $199.12
|
|
|||||
AAH6488714
|
Thermo Scientific Chemicals
H6488714 |
25 g |
Each for $755.42
|
|
|||||
Description
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Chemical Identifiers
4181-05-9 | |
273.34 | |
UESSERYYFWCTBU-UHFFFAOYSA-N | |
77846 | |
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O |
C19H15NO | |
MFCD00145131 | |
4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde | |
4-(N-phenylanilino)benzaldehyde |
Specifications
4181-05-9 | |
Yellow | |
C19H15NO | |
1 g | |
Air sensitive | |
Soluble in methanol. | |
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O | |
273.34 | |
273.33 | |
Solid |
129°C to 133°C (literature) | |
Odorless | |
MFCD00145131 | |
2732795 | |
4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde | |
UESSERYYFWCTBU-UHFFFAOYSA-N | |
4-(N-phenylanilino)benzaldehyde | |
77846 | |
98% | |
4-(Diphenylamino)benzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only