Learn More
Thermo Scientific Chemicals 4-Epichlortetracycline hydrochloride, can be used as secondary standard
4-Epichlortetracycline hydrochloride, CAS # 14297-93-9, is a broad-spectrum, bacteriostatic antibiotic of the tetracycline antibiotic family.
Supplier: Thermo Scientific Chemicals 268235000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• 4-Epichlortetracycline is the hydrochloride salt of 4-Epichlortetracycline, a tetracycline ring structure with a chloride substitution
• 4-Epichlortetracycline hydrochloride (4-Epi-chlortetracycline) is an epimer of chlortetracycline
• 4-Epichlortetracycline suppresses neutrophil activity and dampens the immune response
• Acts as an aminoacyl-tRNA analog to disrupt polypeptide synthesis, reducing bacterial growth Applications
• Can be used for in vitro research on cancer and inflammatory mechanisms
• Intended for use as a selective agent and secondary standard
Specifications
4-Epichlortetracycline hydrochloride | |
101342-45-4 | |
100.0 | |
Orange to Yellow | |
80% min. (as is,calculated) | |
C22H23ClN2O8·ClH | |
500 mg | |
Solubility in water: 67g/L. Other solubilities: 20g/L methanol | |
CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl | |
515.34 | |
515.34 | |
Powder |
97% | |
96.0 | |
250.0°C | |
Authentic | |
Glass bottle | |
MFCD09842537 | |
epichlortetracycline hydrochloride | |
QYAPHLRPFNSDNH-PNEIEYGESA-N | |
(4R,4aS,5aS,6R,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride | |
92043146 | |
≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC) |
Chemical Identifiers
101342-45-4 | |
515.34 | |
QYAPHLRPFNSDNH-PNEIEYGESA-N | |
92043146 | |
CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl |
C22H23ClN2O8·ClH | |
MFCD09842537 | |
epichlortetracycline hydrochloride | |
(4R,4aS,5aS,6R,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Suspected of damaging the unborn child.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o
GHS Signal Word: Warning
RUO – Research Use Only