missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Ethoxyacetophenone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 409590250

 View more versions of this product

Catalog No. AC409590250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Ethoxyacetophenone
98.5
35.0°C to 38.0°C
134°C
Glass bottle
MFCD00009095
4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone
CCOC1=CC=C(C=C1)C(C)=O
164.20
164.2
1676-63-7
100.0
268.0°C to 269.0°C (758.0 mmHg)
98.5% min. (GC)
C10H12O2
25 g
YJFNFQHMQJCPRG-UHFFFAOYSA-N
1-(4-ethoxyphenyl)ethanone
72872
99%

Chemical Identifiers

1676-63-7
164.20
YJFNFQHMQJCPRG-UHFFFAOYSA-N
72872
CCOC1=CC=C(C=C1)C(C)=O
C10H12O2
MFCD00009095
4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone
1-(4-ethoxyphenyl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-825-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only