missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethoxycarbonylphenylboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 367790050

 View more versions of this product

Catalog No. AC367790050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Ethoxycarbonylphenylboronic acid
96.0
184°C to 189°C
Authentic
Glass bottle
MFCD02179441
4-ethoxycarbonylphenyl boronic acid, 4-ethoxycarbonyl phenylboronic acid, 4-ethoxycarbonyl phenyl boronic acid, 4-ethoxycarbonyl benzeneboronic acid, 4-ethoxycarbonylbenzeneboronic acid, ethyl 4-boronobenzoate, p-ethoxycarbonylphenylboronic acid, ethyl 4-dihydroxyboranyl benzoate, 4-carbethoxyphenylboronic acid, benzoic acid, 4-borono-, 1-ethyl ester
CCOC(=O)C1=CC=C(C=C1)B(O)O
193.99
193.99
Powder
4334-88-7
100.0
White
97%
C9H11BO4
5 g
ZLNFACCFYUFTLD-UHFFFAOYSA-N
[4-(ethoxycarbonyl)phenyl]boronic acid
2734350
97%

Chemical Identifiers

4334-88-7
193.99
ZLNFACCFYUFTLD-UHFFFAOYSA-N
2734350
C9H11BO4
MFCD02179441
4-ethoxycarbonylphenyl boronic acid, 4-ethoxycarbonyl phenylboronic acid, 4-ethoxycarbonyl phenyl boronic acid, 4-ethoxycarbonyl benzeneboronic acid, 4-ethoxycarbonylbenzeneboronic acid, ethyl 4-boronobenzoate, p-ethoxycarbonylphenylboronic acid, ethyl 4-dihydroxyboranyl benzoate, 4-carbethoxyphenylboronic acid, benzoic acid, 4-borono-, 1-ethyl ester
CCOC(=O)C1=CC=C(C=C1)B(O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only