missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethyl-4-methylmorpholinium Bromide 97.0+%, TCI America™
Supplier: TCI America E116625G
Specifications
| 4-Ethyl-4-methylmorpholinium Bromide | |
| 177°C | |
| MFCD03095385 | |
| ACQZVWGQFXXTIX-UHFFFAOYSA-M | |
| 4-ethyl-4-methylmorpholin-4-ium;bromide | |
| 103453 | |
| ≥97.0% (T) |
| 65756-41-4 | |
| C7H16BrNO | |
| 25 g | |
| CC[N+]1(CCOCC1)C.[Br-] | |
| 210.115 | |
| 210.12 | |
| Crystalline Powder |
Chemical Identifiers
| 65756-41-4 | |
| 210.115 | |
| ACQZVWGQFXXTIX-UHFFFAOYSA-M | |
| 4-ethyl-4-methylmorpholin-4-ium;bromide |
| C7H16BrNO | |
| MFCD03095385 | |
| 103453 | |
| CC[N+]1(CCOCC1)C.[Br-] |
Safety and Handling
EINECSNumber : (5)-5971
TSCA : Yes