missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Ethylamino)benzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 7409-09-8
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00102118
InChI Key SWXFMMWYVSYQGF-UHFFFAOYSA-N
Synonym 4-ethylamino benzoic acid, 4-ethylaminobenzoic acid, benzoic acid,4-ethylamino, maybridge1_002225, p-athylaminobenzoesaure, swxfmmwyvsyqgf-uhfffaoysa-n
PubChem CID 2775249
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB10315CB
SDS View Documents
Thermo Scientific Chemicals
BTB10315CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB10315DA
SDS View Documents
Thermo Scientific Chemicals
BTB10315DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB10315EA
SDS View Documents
Thermo Scientific Chemicals
BTB10315EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7409-09-8
165.19
SWXFMMWYVSYQGF-UHFFFAOYSA-N
2775249
C9H11NO2
MFCD00102118
4-ethylamino benzoic acid, 4-ethylaminobenzoic acid, benzoic acid,4-ethylamino, maybridge1_002225, p-athylaminobenzoesaure, swxfmmwyvsyqgf-uhfffaoysa-n

Specifications

7409-09-8
100.0
Amber glass bottle
CH3CH2NHC6H4CO2H
250mg
SWXFMMWYVSYQGF-UHFFFAOYSA-N
2775249
97%
96.5
97%
C9H11NO2
MFCD00102118
4-ethylamino benzoic acid, 4-ethylaminobenzoic acid, benzoic acid,4-ethylamino, maybridge1_002225, p-athylaminobenzoesaure, swxfmmwyvsyqgf-uhfffaoysa-n
165.19
165.19
4-(Ethylamino)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'