Learn More
4-Ethylbenzoic acid, 97%
CAS: 619-64-7 | C9H10O2 | 150.18 g/mol
$85.29 - $85.29
Chemical Identifiers
| CAS | 619-64-7 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.18 |
| MDL Number | MFCD00002570 |
| InChI Key | ZQVKTHRQIXSMGY-UHFFFAOYSA-N |
| Synonym | benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid; |
| PubChem CID | 12086 |
| IUPAC Name | 4-ethylbenzoic acid |
| SMILES | CCC1=CC=C(C=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC184490250
|
Thermo Scientific Chemicals
184490250 |
25 g | Glass bottle |
Each for $85.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 619-64-7 | |
| 150.18 | |
| ZQVKTHRQIXSMGY-UHFFFAOYSA-N | |
| 12086 | |
| CCC1=CC=C(C=C1)C(=O)O |
| C9H10O2 | |
| MFCD00002570 | |
| benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid; | |
| 4-ethylbenzoic acid |
Specifications
| 619-64-7 | |
| 100.0 | |
| White to Beige | |
| 97% | |
| C9H10O2 | |
| MFCD00002570 | |
| 09, 529 | |
| Solubility in water: slightly soluble | |
| CCC1=CC=C(C=C1)C(=O)O | |
| 150.18 | |
| 150.18 | |
| Powder |
| 96.0 | |
| 110.0°C to 114.0°C | |
| Authentic | |
| Glass bottle | |
| C2H5C6H4CO2H | |
| 25 g | |
| benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid; | |
| ZQVKTHRQIXSMGY-UHFFFAOYSA-N | |
| 4-ethylbenzoic acid | |
| 12086 | |
| 97% | |
| 4-Ethylbenzoic acid, 97% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 210-605-3
RUO – Research Use Only