missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylheptane 99.0+%, TCI America™

Supplier:  TCI America E02601ML

Catalog No. E02601ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Ethylheptane
Colorless
C9H20
1 mL
XMROPFQWHHUFFS-UHFFFAOYSA-N
4-ethylheptane
16663
≥99.0% (GC)
2216-32-2
141°C
MFCD00048743
1920
CCCC(CC)CCC
128.259
128.26
Liquid

Chemical Identifiers

2216-32-2
128.259
XMROPFQWHHUFFS-UHFFFAOYSA-N
4-ethylheptane
C9H20
MFCD00048743
16663
CCCC(CC)CCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0009

TSCA : No

SDS
Documents

Documents

Promotions

Promotions