missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Ethylphenol, 97%, Thermo Scientific Chemicals

$50.77 - $114.31

Chemical Identifiers

CAS 123-07-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
InChI Key HXDOZKJGKXYMEW-UHFFFAOYSA-N
Synonym p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol
PubChem CID 31242
ChEBI CHEBI:49584
IUPAC Name 4-ethylphenol
SMILES CCC1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC118820050
SDS View Documents
Thermo Scientific Chemicals
118820050
5 g Plastic bottle
Each for $50.77
 
AC118822500
SDS View Documents
Thermo Scientific Chemicals
118822500
250 g Plastic bottle
Each for $114.31
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-07-9
122.17
p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol
CHEBI:49584
CCC1=CC=C(C=C1)O
C8H10O
HXDOZKJGKXYMEW-UHFFFAOYSA-N
31242
4-ethylphenol

Specifications

123-07-9
100.0
Brown to White
100°C
96% min. (GC)
C8H10O
5 g
p-ethylphenol, phenol, 4-ethyl, 1-ethyl-4-hydroxybenzene, phenol, p-ethyl, 1-hydroxy-4-ethylbenzene, para-ethylphenol, 4-hydroxyphenylethane, 4-ethyl-phenol, 4-hydroxyethylbenzene, paraethylphenol
HXDOZKJGKXYMEW-UHFFFAOYSA-N
4-ethylphenol
31242
122.17
Crystalline Mass or Crystals
96.0
41.0°C to 46.0°C
218.0°C to 219.0°C
Authentic
Plastic bottle
C2H5C6H4OH
06,472
Solubility in water: 4900mg/l (25°C). Other solubilities: soluble in alcohol and ether
CCC1=CC=C(C=C1)O
122.17
CHEBI:49584
97%
4-Ethylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-598-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only