Learn More
4-Ethylpyridine, 98%, Thermo Scientific Chemicals
$99.51 - $364.84
Chemical Identifiers
CAS | 536-75-4 |
---|---|
Molecular Formula | C7H9N |
Molecular Weight (g/mol) | 107.156 |
MDL Number | MFCD00006450 |
InChI Key | VJXRKZJMGVSXPX-UHFFFAOYSA-N |
Synonym | pyridine, 4-ethyl, .gamma.-ethylpyridine, 4-ethyl pyridine, unii-3kw0se2tl5, 4-ethyl-pyridine, pyridine, c1-3-alkyl derivs., 3kw0se2tl5, gamma-ethylpyridine, c1-c3 alkylpyridine, 4ethylpyridine |
PubChem CID | 10822 |
IUPAC Name | 4-ethylpyridine |
SMILES | CCC1=CC=NC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1288318
|
Thermo Scientific Chemicals
A1288318 |
50 g |
Each for $99.51
|
|
|||||
AAA1288330
|
Thermo Scientific Chemicals
A1288330 |
250 g |
Each for $364.84
|
|
|||||
Description
4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.
Solubility
Slightly soluble in water (34 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
536-75-4 | |
107.156 | |
VJXRKZJMGVSXPX-UHFFFAOYSA-N | |
10822 | |
CCC1=CC=NC=C1 |
C7H9N | |
MFCD00006450 | |
pyridine, 4-ethyl, .gamma.-ethylpyridine, 4-ethyl pyridine, unii-3kw0se2tl5, 4-ethyl-pyridine, pyridine, c1-3-alkyl derivs., 3kw0se2tl5, gamma-ethylpyridine, c1-c3 alkylpyridine, 4ethylpyridine | |
4-ethylpyridine |
Specifications
536-75-4 | |
0.942 | |
167°C to 168°C | |
Unpleasant | |
1.501 | |
50 g | |
106481 | |
pyridine, 4-ethyl, .gamma.-ethylpyridine, 4-ethyl pyridine, unii-3kw0se2tl5, 4-ethyl-pyridine, pyridine, c1-3-alkyl derivs., 3kw0se2tl5, gamma-ethylpyridine, c1-c3 alkylpyridine, 4ethylpyridine | |
VJXRKZJMGVSXPX-UHFFFAOYSA-N | |
4-ethylpyridine | |
10822 | |
98% |
-91°C | |
10 | |
52°C (126°F) | |
C7H9N | |
MFCD00006450 | |
UN1993 | |
14,3849 | |
Slightly soluble in water (34g/L at 20°C). | |
CCC1=CC=NC=C1 | |
107.156 | |
107.16 | |
4-Ethylpyridine |
Safety and Handling
GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 208-646-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only