missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Fluoro-2'-hydroxyacetophenone, 98%, Thermo Scientific Chemicals

$117.79 - $423.67

Chemical Identifiers

CAS 1481-27-2
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143203
InChI Key HLTBTUXAMVOKIH-UHFFFAOYSA-N
Synonym 4'-fluoro-2'-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethanone, 4-fluoro-2-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethan-1-one, ethanone, 1-4-fluoro-2-hydroxyphenyl, 1-4-fluoro-2-hydroxy-phenyl ethanone, 1-acetyl-4-fluoro-2-hydroxybenzene, pubchem13252, acmc-1bzss, 4'-fluoro-2'-hydroxy
PubChem CID 2737326
IUPAC Name 1-(4-fluoro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(F)C=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1603006
View Documents
Thermo Scientific Chemicals
A1603006
5 g
Each for $117.79
 
AAA1603014
View Documents
Thermo Scientific Chemicals
A1603014
25 g
Each for $423.67
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1481-27-2
154.14
HLTBTUXAMVOKIH-UHFFFAOYSA-N
2737326
CC(=O)C1=CC=C(F)C=C1O
C8H7FO2
MFCD00143203
4'-fluoro-2'-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethanone, 4-fluoro-2-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethan-1-one, ethanone, 1-4-fluoro-2-hydroxyphenyl, 1-4-fluoro-2-hydroxy-phenyl ethanone, 1-acetyl-4-fluoro-2-hydroxybenzene, pubchem13252, acmc-1bzss, 4'-fluoro-2'-hydroxy
1-(4-fluoro-2-hydroxyphenyl)ethanone

Specifications

1481-27-2
65°C (2 mmHg)
C8H7FO2
5 g
4'-fluoro-2'-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethanone, 4-fluoro-2-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethan-1-one, ethanone, 1-4-fluoro-2-hydroxyphenyl, 1-4-fluoro-2-hydroxy-phenyl ethanone, 1-acetyl-4-fluoro-2-hydroxybenzene, pubchem13252, acmc-1bzss, 4'-fluoro-2'-hydroxy
CC(=O)C1=CC=C(F)C=C1O
154.14
154.14
4'-Fluoro-2'-hydroxyacetophenone
29°C to 34°C
98°C (208°F)
MFCD00143203
2249799
HLTBTUXAMVOKIH-UHFFFAOYSA-N
1-(4-fluoro-2-hydroxyphenyl)ethanone
2737326
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only