Learn More
4-Fluoro-2-iodoaniline, 96%, Thermo Scientific Chemicals
$105.92 - $304.97
Chemical Identifiers
CAS | 61272-76-2 |
---|---|
Molecular Formula | C6H5FIN |
Molecular Weight (g/mol) | 237.02 |
MDL Number | MFCD00042584 |
InChI Key | SETOTRGVPANENO-UHFFFAOYSA-N |
Synonym | 2-iodo-4-fluoroaniline, 4-fluoro-2-indoaniline, 4-fluoro-2-iodoanline, 4-fluoro-2-iodo-aniline, 4-fluoro-2-iodophenylamine, 4-fluoro-2-iodo-phenylamine, benzenamine, 4-fluoro-2-iodo, zlchem 368, pubchem3413, 2-iodo-4-fluoro-aniline |
PubChem CID | 2778283 |
IUPAC Name | 4-fluoro-2-iodoaniline |
SMILES | NC1=C(I)C=C(F)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2549906
|
Thermo Scientific Chemicals
B2549906 |
5 g |
Each for $105.92
|
|
|||||
AAB2549914
|
Thermo Scientific Chemicals
B2549914 |
25 g |
Each for $304.97
|
|
|||||
Description
4-Fluoro-2-iodoaniline is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-2-iodoaniline is used as an organic chemical synthesis intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
61272-76-2 | |
237.02 | |
SETOTRGVPANENO-UHFFFAOYSA-N | |
2778283 | |
NC1=C(I)C=C(F)C=C1 |
C6H5FIN | |
MFCD00042584 | |
2-iodo-4-fluoroaniline, 4-fluoro-2-indoaniline, 4-fluoro-2-iodoanline, 4-fluoro-2-iodo-aniline, 4-fluoro-2-iodophenylamine, 4-fluoro-2-iodo-phenylamine, benzenamine, 4-fluoro-2-iodo, zlchem 368, pubchem3413, 2-iodo-4-fluoro-aniline | |
4-fluoro-2-iodoaniline |
Specifications
61272-76-2 | |
MFCD00042584 | |
UN2810 | |
Not miscible in water. | |
NC1=C(I)C=C(F)C=C1 | |
237.02 | |
237.02 | |
4-Fluoro-2-iodoaniline |
C6H5FIN | |
5 g | |
2-iodo-4-fluoroaniline, 4-fluoro-2-indoaniline, 4-fluoro-2-iodoanline, 4-fluoro-2-iodo-aniline, 4-fluoro-2-iodophenylamine, 4-fluoro-2-iodo-phenylamine, benzenamine, 4-fluoro-2-iodo, zlchem 368, pubchem3413, 2-iodo-4-fluoro-aniline | |
SETOTRGVPANENO-UHFFFAOYSA-N | |
4-fluoro-2-iodoaniline | |
2778283 | |
98% |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only