missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-2-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

$165.27 - $165.27

Chemical Identifiers

CAS 450-83-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143318
InChI Key PTKRQIRPNNIORO-UHFFFAOYSA-N
Synonym 2-methoxy-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-2-methoxy, 4-fluoro-2-methoxy-benzaldehyde, 4-fluoro-o-anisaldehyde, fluoromethoxybenzaldehyde4, pubchem2635, 5-fluoro-2-formylanisole, acmc-1ags2, ksc494s3t, 4-fluoro-2-methoxybenzal dehyde
PubChem CID 2774537
IUPAC Name 4-fluoro-2-methoxybenzaldehyde
SMILES COC1=CC(F)=CC=C1C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2609103
View Documents
Thermo Scientific Chemicals
H2609103
1 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

450-83-9
154.14
PTKRQIRPNNIORO-UHFFFAOYSA-N
2774537
COC1=CC(F)=CC=C1C=O
C8H7FO2
MFCD00143318
2-methoxy-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-2-methoxy, 4-fluoro-2-methoxy-benzaldehyde, 4-fluoro-o-anisaldehyde, fluoromethoxybenzaldehyde4, pubchem2635, 5-fluoro-2-formylanisole, acmc-1ags2, ksc494s3t, 4-fluoro-2-methoxybenzal dehyde
4-fluoro-2-methoxybenzaldehyde

Specifications

450-83-9
C8H7FO2
1 g
2-methoxy-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-2-methoxy, 4-fluoro-2-methoxy-benzaldehyde, 4-fluoro-o-anisaldehyde, fluoromethoxybenzaldehyde4, pubchem2635, 5-fluoro-2-formylanisole, acmc-1ags2, ksc494s3t, 4-fluoro-2-methoxybenzal dehyde
COC1=CC(F)=CC=C1C=O
154.14
154.14
4-Fluoro-2-methoxybenzaldehyde
60°C to 62°C
MFCD00143318
Air Sensitive
PTKRQIRPNNIORO-UHFFFAOYSA-N
4-fluoro-2-methoxybenzaldehyde
2774537
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only