Learn More
4-Fluoro-2-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
$55.37 - $181.52
Chemical Identifiers
CAS | 7079-48-3 |
---|---|
Molecular Formula | C7H6ClFO2S |
Molecular Weight (g/mol) | 208.631 |
MDL Number | MFCD03094390 |
InChI Key | XLPGWKNCWMFHOD-UHFFFAOYSA-N |
Synonym | 4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl |
PubChem CID | 2778602 |
IUPAC Name | 4-fluoro-2-methylbenzenesulfonyl chloride |
SMILES | CC1=C(C=CC(=C1)F)S(=O)(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1951703
|
Thermo Scientific Chemicals
L1951703 |
1 g |
Each for $55.37
|
|
|||||
AAL1951706
|
Thermo Scientific Chemicals
L1951706 |
5 g |
Each for $181.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7079-48-3 | |
208.631 | |
XLPGWKNCWMFHOD-UHFFFAOYSA-N | |
2778602 | |
CC1=C(C=CC(=C1)F)S(=O)(=O)Cl |
C7H6ClFO2S | |
MFCD03094390 | |
4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl | |
4-fluoro-2-methylbenzenesulfonyl chloride |
Specifications
7079-48-3 | |
245°C to 246°C | |
C7H6ClFO2S | |
MFCD03094390 | |
UN3265 | |
Moisture sensitive | |
XLPGWKNCWMFHOD-UHFFFAOYSA-N | |
4-fluoro-2-methylbenzenesulfonyl chloride | |
2778602 | |
97% |
1.433 | |
>110°C (230°F) | |
1.537 | |
1 g | |
6720115 | |
4-fluoro-2-methylbenzene-1-sulfonyl chloride, 4-fluoro-2-methylbenzenesulfonylchloride, 4-fluoro-2-methylbenzenesulphonyl chloride, 4-fluoro-2-methyl-benzenesulfonyl chloride, chloro 4-fluoro-2-methylphenyl sulfone, pubchem11719, ablock ab-12-8862, 4-fluoro-2-methylphenylsulfonyl chloride, 4-fluoro-2-methylbenzene-1-sulfonylchloride, benzenesulfonylchloride, 4-fluoro-2-methyl | |
CC1=C(C=CC(=C1)F)S(=O)(=O)Cl | |
208.631 | |
208.64 | |
4-Fluoro-2-methylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only