missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-2-methylbenzoic Acid 98.0+%, TCI America™
$179.73 - $714.95
Chemical Identifiers
CAS | 321-21-1 |
---|---|
Molecular Formula | C8H7FO2 |
Molecular Weight (g/mol) | 154.14 |
MDL Number | MFCD04115880 |
InChI Key | KDXOONIQRUZGSY-UHFFFAOYSA-N |
Synonym | 4-fluoro-o-toluic acid, 2-methyl-4-fluorobenzoic acid, 4-fluoro-2-methylbenzoicacid, 4-fluoro-2-methyl-benzoic acid, benzoic acid, 4-fluoro-2-methyl, pubchem1327, 2-carboxy-5-fluorotoluene, acmc-209hr6, 4-fluoro-2-methyl benzoicacid, rarechem al be 0506 |
PubChem CID | 3860639 |
IUPAC Name | 4-fluoro-2-methylbenzoic acid |
SMILES | CC1=C(C=CC(=C1)F)C(=O)O |
Chemical Identifiers
321-21-1 | |
154.14 | |
KDXOONIQRUZGSY-UHFFFAOYSA-N | |
3860639 | |
CC1=C(C=CC(=C1)F)C(=O)O |
C8H7FO2 | |
MFCD04115880 | |
4-fluoro-o-toluic acid, 2-methyl-4-fluorobenzoic acid, 4-fluoro-2-methylbenzoicacid, 4-fluoro-2-methyl-benzoic acid, benzoic acid, 4-fluoro-2-methyl, pubchem1327, 2-carboxy-5-fluorotoluene, acmc-209hr6, 4-fluoro-2-methyl benzoicacid, rarechem al be 0506 | |
4-fluoro-2-methylbenzoic acid |
Specifications
321-21-1 | |
White-Yellow | |
MFCD04115880 | |
4-fluoro-o-toluic acid, 2-methyl-4-fluorobenzoic acid, 4-fluoro-2-methylbenzoicacid, 4-fluoro-2-methyl-benzoic acid, benzoic acid, 4-fluoro-2-methyl, pubchem1327, 2-carboxy-5-fluorotoluene, acmc-209hr6, 4-fluoro-2-methyl benzoicacid, rarechem al be 0506 | |
CC1=C(C=CC(=C1)F)C(=O)O | |
154.14 | |
154.14 | |
Crystalline Powder |
174°C | |
C8H7FO2 | |
5 g | |
KDXOONIQRUZGSY-UHFFFAOYSA-N | |
4-fluoro-2-methylbenzoic acid | |
3860639 | |
≥98.0% (GC,T) | |
4-Fluoro-2-methylbenzoic Acid |
Safety and Handling
TSCA : No