missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzoyl Chloride 98.0+%, TCI America™

$182.68 - $548.03

Chemical Identifiers

CAS 455-84-5
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
MDL Number MFCD01631426
InChI Key FMFLKTDRPXYUGP-UHFFFAOYSA-N
Synonym 4-fluoro-3-methylbenzoylchloride, 4-fluoro-m-toluoyl chloride, 4-fluoro-3-methyl-benzoyl chloride, benzoyl chloride, 4-fluoro-3-methyl, acmc-1arwr, 5-chlorocarbonyl-2-fluorotoluene, 4-fluoranyl-3-methyl-benzoyl chloride, 4-fluoro-3-methyl benzoyl chloride, 4-fluoro-3-methyl-benzoic acid chloride, 4-fluoro-3-methylbenzoyl chloride
PubChem CID 2774589
IUPAC Name 4-fluoro-3-methylbenzoyl chloride
SMILES CC1=C(C=CC(=C1)C(=O)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F11165G
View Documents
TCI America
F11165G
5 g N/A
Only null left
 
F111625G
View Documents
TCI America
F111625G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

455-84-5
172.583
FMFLKTDRPXYUGP-UHFFFAOYSA-N
2774589
CC1=C(C=CC(=C1)C(=O)Cl)F
C8H6ClFO
MFCD01631426
4-fluoro-3-methylbenzoylchloride, 4-fluoro-m-toluoyl chloride, 4-fluoro-3-methyl-benzoyl chloride, benzoyl chloride, 4-fluoro-3-methyl, acmc-1arwr, 5-chlorocarbonyl-2-fluorotoluene, 4-fluoranyl-3-methyl-benzoyl chloride, 4-fluoro-3-methyl benzoyl chloride, 4-fluoro-3-methyl-benzoic acid chloride, 4-fluoro-3-methylbenzoyl chloride
4-fluoro-3-methylbenzoyl chloride

Specifications

455-84-5
115°C
MFCD01631426
3265
FMFLKTDRPXYUGP-UHFFFAOYSA-N
4-fluoro-3-methylbenzoyl chloride
2774589
≥98.0% (GC,T)
4-Fluoro-3-methylbenzoyl Chloride
Yellow
C8H6ClFO
5 g
4-fluoro-3-methylbenzoylchloride, 4-fluoro-m-toluoyl chloride, 4-fluoro-3-methyl-benzoyl chloride, benzoyl chloride, 4-fluoro-3-methyl, acmc-1arwr, 5-chlorocarbonyl-2-fluorotoluene, 4-fluoranyl-3-methyl-benzoyl chloride, 4-fluoro-3-methyl benzoyl chloride, 4-fluoro-3-methyl-benzoic acid chloride, 4-fluoro-3-methylbenzoyl chloride
CC1=C(C=CC(=C1)C(=O)Cl)F
172.583
172.58
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'