missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Fluoro-3'-nitroacetophenone, 98%, Thermo Scientific™

$110.74 - $110.74

Chemical Identifiers

CAS 400-93-1
Molecular Formula C8H6FNO3
Molecular Weight (g/mol) 183.14
MDL Number MFCD00115369
InChI Key PTCNZDJJIOLIKQ-UHFFFAOYSA-N
Synonym 4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone
PubChem CID 2734612
IUPAC Name 1-(4-fluoro-3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366010050
SDS View Documents
Thermo Scientific Chemicals
366010050
5 g Glass bottle N/A
 
Description

Description

Specifications

Chemical Identifiers

400-93-1
183.14
PTCNZDJJIOLIKQ-UHFFFAOYSA-N
2734612
CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]
C8H6FNO3
MFCD00115369
4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone
1-(4-fluoro-3-nitrophenyl)ethanone

Specifications

400-93-1
100.0
Yellow
97.5% min. (GC)
C8H6FNO3
5 g
PTCNZDJJIOLIKQ-UHFFFAOYSA-N
1-(4-fluoro-3-nitrophenyl)ethanone
2734612
98%
4'-Fluoro-3'-nitroacetophenone
97.5
46°C to 50°C
Authentic
Glass bottle
MFCD00115369
4'-fluoro-3'-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethanone, 4-fluoro-3-nitroacetophenone, 1-4-fluoro-3-nitrophenyl ethan-1-one, 1-4-fluoro-3-nitro-phenyl ethanone, 4-fluoro-3'-nitroacetophenone, ethanone, 1-4-fluoro-3-nitrophenyl, 4-fluoro-3-nitroacetophneone, 3'-nitro-4'-fluoroacetophenone
CC(=O)C1=CC(=C(C=C1)F)[N+](=O)[O-]
183.14
183.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only