missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-nitrobenzaldehyde 95.0+%, TCI America™

$43.25 - $122.84

Chemical Identifiers

CAS 42564-51-2
Molecular Formula C7H4FNO3
Molecular Weight (g/mol) 169.111
MDL Number MFCD01861388
InChI Key ILKWFRCNNILIJW-UHFFFAOYSA-N
Synonym 3-nitro-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzaldehyde, pubchem22572, acmc-1ao2q, 3-nitro-4 fluorobenzaldehyde, ksc497m9r, 2-fluoro-5-formylnitrobenzene, #, 4-fluoranyl-3-nitro-benzaldehyde
PubChem CID 598129
IUPAC Name 4-fluoro-3-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)[N+](=O)[O-])F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F09561G
View Documents
TCI America
F09561G
1 g
Each for $43.25
 
F09565G
View Documents
TCI America
F09565G
5 g
Each for $122.84
 
Description

Description

Specifications

Chemical Identifiers

42564-51-2
169.111
ILKWFRCNNILIJW-UHFFFAOYSA-N
598129
C1=CC(=C(C=C1C=O)[N+](=O)[O-])F
C7H4FNO3
MFCD01861388
3-nitro-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzaldehyde, pubchem22572, acmc-1ao2q, 3-nitro-4 fluorobenzaldehyde, ksc497m9r, 2-fluoro-5-formylnitrobenzene, #, 4-fluoranyl-3-nitro-benzaldehyde
4-fluoro-3-nitrobenzaldehyde

Specifications

42564-51-2
Yellow
MFCD01861388
3-nitro-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzaldehyde, pubchem22572, acmc-1ao2q, 3-nitro-4 fluorobenzaldehyde, ksc497m9r, 2-fluoro-5-formylnitrobenzene, #, 4-fluoranyl-3-nitro-benzaldehyde
C1=CC(=C(C=C1C=O)[N+](=O)[O-])F
169.111
169.11
Crystalline Powder
45°C
C7H4FNO3
1 g
ILKWFRCNNILIJW-UHFFFAOYSA-N
4-fluoro-3-nitrobenzaldehyde
598129
≥95.0% (GC)
4-Fluoro-3-nitrobenzaldehyde
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'