missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals
$69.15 - $662.08
Chemical Identifiers
CAS | 169243-86-1 |
---|---|
Molecular Formula | C24H20FNO4 |
Molecular Weight (g/mol) | 405.43 |
MDL Number | MFCD00191197 |
InChI Key | IXUMACXMEZBPJG-QFIPXVFZSA-N |
Synonym | fmoc-4-fluoro-l-phenylalanine, fmoc-l-4-fluorophenylalanine, fmoc-phe 4-f-oh, fmoc-l-4-fluorophe, fmoc-4-fluoro-l-phe, fmoc-4-fluoro-l-phe-oh, fmoc-l-phe 4-f-oh, fmoc-p-fluoro-phe-oh, l-4-fluoro-phenyl-n-fmoc-alanine, s-n-fmoc-4-fluorophenylalanine |
PubChem CID | 2734466 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-fluorophenyl)propanoic acid |
SMILES | OC(=O)[C@H](CC1=CC=C(F)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH51972MD
|
Thermo Scientific Chemicals
H51972MD |
250 mg |
Each for $69.15
|
|
|||||
AAH5197203
|
Thermo Scientific Chemicals
H5197203 |
1 g |
Each for $168.67
|
|
|||||
AAH5197206
|
Thermo Scientific Chemicals
H5197206 |
5 g |
Each for $662.08
|
|
|||||
Description
4-Fluoro-N-Fmoc-L-phenylalanine derivative was introduced in collaboration with Yu and coworkers in reflection to a methodology reported in C-H Activation. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-N-Fmoc-L-phenylalanine derivative was introduced in collaboration with Yu and coworkers in reflection to a methodology reported in C-H Activation. It is also used as pharmaceutical intermediate.
Solubility
Soluble in Dimethylformamide(DMF).
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Chemical Identifiers
169243-86-1 | |
405.43 | |
IXUMACXMEZBPJG-QFIPXVFZSA-N | |
2734466 | |
OC(=O)[C@H](CC1=CC=C(F)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
C24H20FNO4 | |
MFCD00191197 | |
fmoc-4-fluoro-l-phenylalanine, fmoc-l-4-fluorophenylalanine, fmoc-phe 4-f-oh, fmoc-l-4-fluorophe, fmoc-4-fluoro-l-phe, fmoc-4-fluoro-l-phe-oh, fmoc-l-phe 4-f-oh, fmoc-p-fluoro-phe-oh, l-4-fluoro-phenyl-n-fmoc-alanine, s-n-fmoc-4-fluorophenylalanine | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-fluorophenyl)propanoic acid |
Specifications
169243-86-1 | |
MFCD00191197 | |
8287377 | |
Soluble in Dimethylformamide(DMF). | |
OC(=O)[C@H](CC1=CC=C(F)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
405.43 | |
405.42 | |
4-Fluoro-N-Fmoc-L-phenylalanine |
C24H20FNO4 | |
250 mg | |
fmoc-4-fluoro-l-phenylalanine, fmoc-l-4-fluorophenylalanine, fmoc-phe 4-f-oh, fmoc-l-4-fluorophe, fmoc-4-fluoro-l-phe, fmoc-4-fluoro-l-phe-oh, fmoc-l-phe 4-f-oh, fmoc-p-fluoro-phe-oh, l-4-fluoro-phenyl-n-fmoc-alanine, s-n-fmoc-4-fluorophenylalanine | |
IXUMACXMEZBPJG-QFIPXVFZSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-fluorophenyl)propanoic acid | |
2734466 | |
95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only