missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-N,N-dimethyl-2-nitroaniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6335703

 View more versions of this product

Catalog No. AAH6335703


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Fluoro-N,N-dimethyl-2-nitroaniline
C8H9FN2O2
1 g
n,n-dimethyl 4-fluoro-2-nitroaniline, n,n-dimethyl-4-fluoro-2-nitroaniline, benzenamine,4-fluoro-n,n-dimethyl-2-nitro, pubchem3366, acmc-1b82t, 4-fluoro-2-nitrophenyl dimethylamine, 4-fluoranyl-n,n-dimethyl-2-nitro-aniline
CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
184.17
184.17
Solid
69261-01-4
MFCD04116334
UN2811
RGCUVPLVTHHXMT-UHFFFAOYSA-N
4-fluoro-N,N-dimethyl-2-nitroaniline
16440018
97%

Chemical Identifiers

69261-01-4
184.17
RGCUVPLVTHHXMT-UHFFFAOYSA-N
16440018
CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
C8H9FN2O2
MFCD04116334
n,n-dimethyl 4-fluoro-2-nitroaniline, n,n-dimethyl-4-fluoro-2-nitroaniline, benzenamine,4-fluoro-n,n-dimethyl-2-nitro, pubchem3366, acmc-1b82t, 4-fluoro-2-nitrophenyl dimethylamine, 4-fluoranyl-n,n-dimethyl-2-nitro-aniline
4-fluoro-N,N-dimethyl-2-nitroaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions