missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzamidine hydrochloride, 95%, Thermo Scientific™
$85.62 - $340.70
Chemical Identifiers
| CAS | 456-14-4 |
|---|---|
| Molecular Formula | C7H8ClFN2 |
| Molecular Weight (g/mol) | 174.60 |
| MDL Number | MFCD04114421 |
| InChI Key | JQDATBKJKUWNGA-UHFFFAOYSA-N |
| Synonym | 4-fluorobenzamidine hydrochloride, 4-fluorobenzimidamide hydrochloride, 4-fluorobenzamidine hcl, 4-fluoro-benzamidine hydrochloride, 4-fluorobenzenecarboximidamide hydrochloride, 4-fluorobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-fluoro-, monohydrochloride, 4-fluorobenzimidamidehydrochloride, 4-fluorobenzamidine hydrogen chloride, 4-fluoranylbenzenecarboximidamide hydrochloride |
| PubChem CID | 12456160 |
| IUPAC Name | 4-fluorobenzenecarboximidamide;hydrochloride |
| SMILES | [H+].[Cl-].NC(=N)C1=CC=C(F)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6157003
|
Thermo Scientific Chemicals
H6157003 |
1 g |
Each for $85.62
|
|
|||||
|
AAH6157006
|
Thermo Scientific Chemicals
H6157006 |
5 g |
Each for $340.70
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-Fluorobenzamidine hydrochloride is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 456-14-4 | |
| 174.60 | |
| JQDATBKJKUWNGA-UHFFFAOYSA-N | |
| 12456160 | |
| [H+].[Cl-].NC(=N)C1=CC=C(F)C=C1 |
| C7H8ClFN2 | |
| MFCD04114421 | |
| 4-fluorobenzamidine hydrochloride, 4-fluorobenzimidamide hydrochloride, 4-fluorobenzamidine hcl, 4-fluoro-benzamidine hydrochloride, 4-fluorobenzenecarboximidamide hydrochloride, 4-fluorobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-fluoro-, monohydrochloride, 4-fluorobenzimidamidehydrochloride, 4-fluorobenzamidine hydrogen chloride, 4-fluoranylbenzenecarboximidamide hydrochloride | |
| 4-fluorobenzenecarboximidamide;hydrochloride |
Specifications
| 456-14-4 | |
| MFCD04114421 | |
| 4-fluorobenzamidine hydrochloride, 4-fluorobenzimidamide hydrochloride, 4-fluorobenzamidine hcl, 4-fluoro-benzamidine hydrochloride, 4-fluorobenzenecarboximidamide hydrochloride, 4-fluorobenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-fluoro-, monohydrochloride, 4-fluorobenzimidamidehydrochloride, 4-fluorobenzamidine hydrogen chloride, 4-fluoranylbenzenecarboximidamide hydrochloride | |
| [H+].[Cl-].NC(=N)C1=CC=C(F)C=C1 | |
| 174.60 | |
| 174.6 | |
| Solid |
| C7H8ClFN2 | |
| 1 g | |
| JQDATBKJKUWNGA-UHFFFAOYSA-N | |
| 4-fluorobenzenecarboximidamide;hydrochloride | |
| 12456160 | |
| 95% | |
| 4-Fluorobenzamidine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures