Learn More
4-Fluorobenzyl chloride, 99%
CAS: 352-11-4 | C7H6ClF | 144.573 g/mol
$66.56 - $614.33
Chemical Identifiers
| CAS | 352-11-4 |
|---|---|
| Molecular Formula | C7H6ClF |
| Molecular Weight (g/mol) | 144.573 |
| MDL Number | MFCD00000913 |
| InChI Key | IZXWCDITFDNEBY-UHFFFAOYSA-N |
| Synonym | 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene |
| PubChem CID | 9602 |
| IUPAC Name | 1-(chloromethyl)-4-fluorobenzene |
| SMILES | C1=CC(=CC=C1CCl)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1468614
|
Thermo Scientific Chemicals
A1468614 |
25 g |
Each for $66.56
|
|
|||||
|
AAA1468622
|
Thermo Scientific Chemicals
A1468622 |
100 g |
Each for $171.59
|
|
|||||
|
AAA1468636
|
Thermo Scientific Chemicals
A1468636 |
500 g |
Each for $614.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 352-11-4 | |
| 144.573 | |
| IZXWCDITFDNEBY-UHFFFAOYSA-N | |
| 9602 | |
| C1=CC(=CC=C1CCl)F |
| C7H6ClF | |
| MFCD00000913 | |
| 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene | |
| 1-(chloromethyl)-4-fluorobenzene |
Specifications
| 352-11-4 | |
| 1.214 | |
| 69°C (156°F) | |
| C7H6ClF | |
| MFCD00000913 | |
| UN3265 | |
| 4-fluorobenzyl chloride, 1-chloromethyl-4-fluorobenzene, alpha-chloro-p-fluorotoluene, p-fluorobenzyl chloride, 4-fluorobenzylchloride, benzene, 1-chloromethyl-4-fluoro, 1-chloromethyl-4-fluoro-benzene, alpha-chloro-4-fluorotoluene, ccris 5107, chloro-4-fluorotoluene | |
| C1=CC(=CC=C1CCl)F | |
| 144.573 | |
| 144.58 | |
| 4-Fluorobenzyl chloride |
| -18°C | |
| 185°C | |
| Pungent | |
| 1.513 | |
| 25 g | |
| 742272 | |
| IZXWCDITFDNEBY-UHFFFAOYSA-N | |
| 1-(chloromethyl)-4-fluorobenzene | |
| 9602 | |
| 99% |
Safety and Handling
GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 206-516-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only