missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific™

Chemical Identifiers

CAS 352-13-6
Molecular Formula C6H4BrFMg
Molecular Weight (g/mol) 199.305
MDL Number MFCD00009667
InChI Key BRKADVNLTRCLOW-UHFFFAOYSA-M
Synonym 4-fluorophenylmagnesium bromide, grignard reagent, bromo 4-fluorophenyl magnesium, bromo 4-fluorophenyl magnesium, 4-fluorophenyl magnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-flourophenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide
PubChem CID 2734897
IUPAC Name magnesium;fluorobenzene;bromide
SMILES C1=CC(=CC=[C-]1)F.[Mg+2].[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC377371000
SDS View Documents
Thermo Scientific Chemicals
377371000
100mL AcroSeal™ Glass Bottle in Tin Can N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC377378000
SDS View Documents
Thermo Scientific Chemicals
377378000
800mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

352-13-6
199.305
BRKADVNLTRCLOW-UHFFFAOYSA-M
2734897
C1=CC(=CC=[C-]1)F.[Mg+2].[Br-]
C6H4BrFMg
MFCD00009667
4-fluorophenylmagnesium bromide, grignard reagent, bromo 4-fluorophenyl magnesium, bromo 4-fluorophenyl magnesium, 4-fluorophenyl magnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-flourophenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide
magnesium;fluorobenzene;bromide

Specifications

352-13-6,60-29-7
42.04,61.78
F(C6H4)MgBr
2 to 2.2M
4-fluorophenylmagnesium bromide, grignard reagent, bromo 4-fluorophenyl magnesium, bromo 4-fluorophenyl magnesium, 4-fluorophenyl magnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-flourophenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide
C1=CC(=CC=[C-]1)F.[Mg+2].[Br-]
199.305
199.3
100mL
38.22,57.96
C6H4BrFMg
MFCD00009667
1.0430g/mL
BRKADVNLTRCLOW-UHFFFAOYSA-M
magnesium;fluorobenzene;bromide
2734897
AcroSeal™ Glass Bottle in Tin Can
4-Fluorophenylmagnesium bromide, 2M Solution in Diethyl Ether
Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gas. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. Reacts violently with water.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only