missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Fmoc-aminomethyl)benzoic acid, 95%, Thermo Scientific Chemicals

$127.29 - $410.89

Chemical Identifiers

CAS 164470-64-8
Molecular Formula C23H19NO4
Molecular Weight (g/mol) 373.41
MDL Number MFCD01074693
InChI Key JRHUROPSUJVMNH-UHFFFAOYSA-N
Synonym fmoc-4-amb-oh, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl benzoic acid, 4-9h-fluoren-9-ylmethoxycarbonylamino methyl benzoic acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino methyl benzoic acid, 4-n-fmoc-aminomethyl-benzoic acid, 4-fmoc-aminomethyl benzoicacid, 4-fmoc-aminomethyl benzoic acid, fmoc-4-amb-oh fmoc-4-aminomethyl benzoic acid
PubChem CID 2756083
IUPAC Name 4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]benzoic acid
SMILES OC(=O)C1=CC=C(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6634006
View Documents
Thermo Scientific Chemicals
H6634006
5 g
Each for $127.29
 
AAH6634014
View Documents
Thermo Scientific Chemicals
H6634014
25 g
Each for $410.89
 
Description

Description

4-(Fmoc-aminomethyl)benzoic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Fmoc-aminomethyl)benzoic acid is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Incompatible with heat and oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications

Chemical Identifiers

164470-64-8
373.41
JRHUROPSUJVMNH-UHFFFAOYSA-N
2756083
OC(=O)C1=CC=C(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C=C1
C23H19NO4
MFCD01074693
fmoc-4-amb-oh, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl benzoic acid, 4-9h-fluoren-9-ylmethoxycarbonylamino methyl benzoic acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino methyl benzoic acid, 4-n-fmoc-aminomethyl-benzoic acid, 4-fmoc-aminomethyl benzoicacid, 4-fmoc-aminomethyl benzoic acid, fmoc-4-amb-oh fmoc-4-aminomethyl benzoic acid
4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]benzoic acid

Specifications

164470-64-8
White
MFCD01074693
7391207
Slightly soluble in water.
OC(=O)C1=CC=C(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C=C1
373.41
373.41
Powder
224°C to 228°C
C23H19NO4
5 g
fmoc-4-amb-oh, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl benzoic acid, 4-9h-fluoren-9-ylmethoxycarbonylamino methyl benzoic acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino methyl benzoic acid, 4-n-fmoc-aminomethyl-benzoic acid, 4-fmoc-aminomethyl benzoicacid, 4-fmoc-aminomethyl benzoic acid, fmoc-4-amb-oh fmoc-4-aminomethyl benzoic acid
JRHUROPSUJVMNH-UHFFFAOYSA-N
4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]benzoic acid
2756083
95%
4-(Fmoc-aminomethyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only