Learn More
4-Formylphenoxyacetic acid, 98%, Thermo Scientific Chemicals
$135.66 - $1926.01
Chemical Identifiers
CAS | 22042-71-3 |
---|---|
Molecular Formula | C9H7O4 |
Molecular Weight (g/mol) | 179.15 |
MDL Number | MFCD00016613 |
InChI Key | OYNIIKHNXNPSAG-UHFFFAOYSA-M |
Synonym | 4-formylphenoxyacetic acid, 4-formylphenoxy acetic acid, 2-4-formylphenoxy acetic acid, p-formylphenoxyacetic acid, acetic acid, 4-formylphenoxy, acetic acid, 2-4-formylphenoxy, p-formylphenoxy acetic acid, 4-carboxymethyloxy benzaldehyde, 4-formyl phenoxy acetic acid, 4-formylphenoxyactic acid |
PubChem CID | 89177 |
IUPAC Name | 2-(4-formylphenoxy)acetic acid |
SMILES | [O-]C(=O)COC1=CC=C(C=O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1396009
|
Thermo Scientific Chemicals
A1396009 |
10 g |
Each for $135.66
|
|
|||||
AAA1396018
|
Thermo Scientific Chemicals
A1396018 |
50 g |
Each for $485.77
|
|
|||||
AAA1396030
|
Thermo Scientific Chemicals
A1396030 |
250 g |
Each for $1,926.01
|
|
|||||
Description
4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Formylphenoxyacetic acid is used as an organic chemical synthesis intermediate.
Solubility
Soluble in hot methanol within almost transparency.
Notes
Air sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
22042-71-3 | |
179.15 | |
OYNIIKHNXNPSAG-UHFFFAOYSA-M | |
89177 | |
[O-]C(=O)COC1=CC=C(C=O)C=C1 |
C9H7O4 | |
MFCD00016613 | |
4-formylphenoxyacetic acid, 4-formylphenoxy acetic acid, 2-4-formylphenoxy acetic acid, p-formylphenoxyacetic acid, acetic acid, 4-formylphenoxy, acetic acid, 2-4-formylphenoxy, p-formylphenoxy acetic acid, 4-carboxymethyloxy benzaldehyde, 4-formyl phenoxy acetic acid, 4-formylphenoxyactic acid | |
2-(4-formylphenoxy)acetic acid |
Specifications
22042-71-3 | |
C9H7O4 | |
10 g | |
Air Sensitive | |
Soluble in hot methanol within almost transparency. | |
[O-]C(=O)COC1=CC=C(C=O)C=C1 | |
179.15 | |
180.16 | |
4-Formylphenoxyacetic acid |
193°C to 198°C | |
MFCD00016613 | |
779885 | |
4-formylphenoxyacetic acid, 4-formylphenoxy acetic acid, 2-4-formylphenoxy acetic acid, p-formylphenoxyacetic acid, acetic acid, 4-formylphenoxy, acetic acid, 2-4-formylphenoxy, p-formylphenoxy acetic acid, 4-carboxymethyloxy benzaldehyde, 4-formyl phenoxy acetic acid, 4-formylphenoxyactic acid | |
OYNIIKHNXNPSAG-UHFFFAOYSA-M | |
2-(4-formylphenoxy)acetic acid | |
89177 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 244-749-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only