missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
4-Formylphenylboronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 359380010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
4-Formylphenylboronic acid | |
87199-17-5 | |
100.0 | |
96% min. (HPLC) | |
C7H7BO3 | |
1g | |
VXWBQOJISHAKKM-UHFFFAOYSA-N | |
(4-formylphenyl)boronic acid | |
591073 | |
97% |
97% | |
96.0 | |
Authentic | |
Glass bottle | |
MFCD00151823 | |
4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid | |
OB(O)C1=CC=C(C=O)C=C1 | |
149.94 | |
149.94 |
Identifiants chimiques
87199-17-5 | |
149.94 | |
VXWBQOJISHAKKM-UHFFFAOYSA-N | |
591073 | |
OB(O)C1=CC=C(C=O)C=C1 |
C7H7BO3 | |
MFCD00151823 | |
4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid | |
(4-formylphenyl)boronic acid |
Sécurité et manipulation
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C
RUO â Research Use Only