Learn More
4-Formylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 445040050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Formylphenylboronic acid pinacol ester | |
59.0°C to 63.0°C | |
C13H17BO3 | |
5 g | |
DMBMXJJGPXADPO-UHFFFAOYSA-N | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde | |
2769536 | |
97% |
128376-64-7 | |
96% min. (GC) | |
MFCD04972375 | |
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylphenylboronic acid pinacol ester, 4-formylbenzeneboronic acid, pinacol ester, 4-formylphenylboronic acid pinacolate, 4-formylphenylboronic acid pinacol cyclic ester, 4-formylphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylbenzeneboronic acid pinacol ester, benzaldehyde, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-formylphenylboronic acid, pinacol cyclic ester | |
CC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1 | |
232.09 | |
232.09 |
Chemical Identifiers
128376-64-7 | |
232.09 | |
DMBMXJJGPXADPO-UHFFFAOYSA-N | |
2769536 | |
CC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1 |
C13H17BO3 | |
MFCD04972375 | |
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylphenylboronic acid pinacol ester, 4-formylbenzeneboronic acid, pinacol ester, 4-formylphenylboronic acid pinacolate, 4-formylphenylboronic acid pinacol cyclic ester, 4-formylphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylbenzeneboronic acid pinacol ester, benzaldehyde, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-formylphenylboronic acid, pinacol cyclic ester | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only