missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Heptanone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 146555000

 View more versions of this product

Catalog No. AC146555000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Heptanone
123-19-3
100.0
Colorless to Yellow
145.0°C
Authentic
Glass bottle
1.4060 to 1.4080
MFCD00009403
01,699
15,3386
Solubility in water: 4.6g/L (20°C). Other solubilities: soluble in alcohol and ether
CCCC(=O)CCC
114.19
0.73 mPa.s (20°C)
98%
98%
97.5
-33.0°C
0.8100g/mL
48°C
97.5% min. (GC)
C7H14O
CH3CH2CH2COCH2CH2CH3
500 mL
0.81
4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon
HCFAJYNVAYBARA-UHFFFAOYSA-N
heptan-4-one
31246
114.19
Liquid

Chemical Identifiers

123-19-3
114.19
HCFAJYNVAYBARA-UHFFFAOYSA-N
31246
CCCC(=O)CCC
C7H14O
MFCD00009403
4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon
heptan-4-one
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-608-9

RTECSNumber : MJ5600000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only