Learn More
4-Hydrazinobenzoic acid hydrochloride, 98%, Thermo Scientific Chemicals
$74.29 - $74.29
Chemical Identifiers
CAS | 24589-77-3 |
---|---|
Molecular Formula | C7H9ClN2O2 |
Molecular Weight (g/mol) | 188.61 |
MDL Number | MFCD00039073 |
InChI Key | XHLQMKQBCHYRLC-UHFFFAOYSA-N |
Synonym | 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride |
PubChem CID | 90547 |
IUPAC Name | 4-hydrazinylbenzoic acid;hydrochloride |
SMILES | [H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2032609
|
Thermo Scientific Chemicals
B2032609 |
10 g |
N/A
|
|
|||||
Description
4-Hydrazinobenzoic acid hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydrazinobenzoic acid hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It may be used for the preparation of 4-(3-(2-(tert-butoxycarbonylamino)ethyl)-5-hydroxy-1H-pyrazol-1-yl)benzoic acid.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
24589-77-3 | |
188.61 | |
XHLQMKQBCHYRLC-UHFFFAOYSA-N | |
90547 | |
[H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O |
C7H9ClN2O2 | |
MFCD00039073 | |
4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride | |
4-hydrazinylbenzoic acid;hydrochloride |
Specifications
24589-77-3 | |
C7H9ClN2O2 | |
10 g | |
4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride | |
XHLQMKQBCHYRLC-UHFFFAOYSA-N | |
4-hydrazinylbenzoic acid;hydrochloride | |
90547 | |
98% |
252°C to 254°C (decomposition) | |
MFCD00039073 | |
Hygroscopic | |
Soluble in water. | |
[H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O | |
188.61 | |
188.61 | |
4-Hydrazinobenzoic acid hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-330-0
RTECSNumber : DH1700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only