missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-1-indanone, 97%, Thermo Scientific™

$170.05 - $668.23

Chemical Identifiers

CAS 40731-98-4
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00143330
InChI Key CKSCMRNFDBWFND-UHFFFAOYSA-N
Synonym 4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659
PubChem CID 590547
IUPAC Name 4-hydroxy-2,3-dihydroinden-1-one
SMILES OC1=CC=CC2=C1CCC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5618403
View Documents
Thermo Scientific Chemicals
H5618403
1 g
Each for $170.05
 
AAH5618406
View Documents
Thermo Scientific Chemicals
H5618406
5 g
Each for $668.23
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Hydroxy-1-indanone is a hydroxylated indanone used for synthesis. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in ethanol. (25mg/ml)

Notes
Incompatible with oxidizing agents. Store at room temperature. Avoid excessive heat and light.
Specifications

Chemical Identifiers

40731-98-4
148.16
CKSCMRNFDBWFND-UHFFFAOYSA-N
590547
OC1=CC=CC2=C1CCC2=O
C9H8O2
MFCD00143330
4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659
4-hydroxy-2,3-dihydroinden-1-one

Specifications

40731-98-4
C9H8O2
1 g
Soluble in ethanol. (25mg/ml)
OC1=CC=CC2=C1CCC2=O
148.16
148.16
4-Hydroxy-1-indanone
242°C to 244°C
MFCD00143330
4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659
CKSCMRNFDBWFND-UHFFFAOYSA-N
4-hydroxy-2,3-dihydroinden-1-one
590547
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'