missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hydroxy-2(5H)-furanone, 95%, Thermo Scientific Chemicals
$107.74 - $107.74
Chemical Identifiers
CAS | 541-57-1 |
---|---|
Molecular Formula | C4H4O3 |
Molecular Weight (g/mol) | 100.07 |
MDL Number | MFCD00052187 |
InChI Key | JZQBAGOECGRTSA-UHFFFAOYSA-N |
Synonym | 4-hydroxyfuran-2 5h-one, 4-hydroxy-2,5-dihydrofuran-2-one, 4-hydroxy-2 5h-furanone, 2 5h-furanone, 4-hydroxy, 4-hydroxy-5h-furan-2-one, 5-hydroxyfuran-3-one, 5-hydroxyfuran-3 2h-one, pubchem21770, acmc-209xna, tetronic acid 5g |
PubChem CID | 54683813 |
IUPAC Name | 3-hydroxy-2H-furan-5-one |
SMILES | OC1=CC(=O)OC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC235260010
|
Thermo Scientific Chemicals
235260010 |
1 g | Glass bottle |
Each for $107.74
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
541-57-1 | |
100.07 | |
JZQBAGOECGRTSA-UHFFFAOYSA-N | |
54683813 | |
OC1=CC(=O)OC1 |
C4H4O3 | |
MFCD00052187 | |
4-hydroxyfuran-2 5h-one, 4-hydroxy-2,5-dihydrofuran-2-one, 4-hydroxy-2 5h-furanone, 2 5h-furanone, 4-hydroxy, 4-hydroxy-5h-furan-2-one, 5-hydroxyfuran-3-one, 5-hydroxyfuran-3 2h-one, pubchem21770, acmc-209xna, tetronic acid 5g | |
3-hydroxy-2H-furan-5-one |
Specifications
541-57-1 | |
100.0 | |
Beige-Yellow | |
94% min. (HPLC) | |
C4H4O3 | |
1 g | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,acetonitrile,dmso | |
OC1=CC(=O)OC1 | |
100.07 | |
100.07 | |
Crystalline Powder |
94.0 | |
135°C to 137°C | |
Authentic | |
Glass bottle | |
MFCD00052187 | |
4-hydroxyfuran-2 5h-one, 4-hydroxy-2,5-dihydrofuran-2-one, 4-hydroxy-2 5h-furanone, 2 5h-furanone, 4-hydroxy, 4-hydroxy-5h-furan-2-one, 5-hydroxyfuran-3-one, 5-hydroxyfuran-3 2h-one, pubchem21770, acmc-209xna, tetronic acid 5g | |
JZQBAGOECGRTSA-UHFFFAOYSA-N | |
3-hydroxy-2H-furan-5-one | |
54683813 | |
95% | |
4-Hydroxy-2(5H)-furanone |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
EINECSNumber : 208-785-3
RUO – Research Use Only