missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-2-butanone, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1145614

 View more versions of this product

Catalog No. AAL1145614

Explore available promotions

Description

Description

4-Hydroxy-2-butanone is used in the preparation of fused benzazepine molecules as selective D3 receptor antagonists with pharmaceutical activity. It is also used in the preparation of verrucarin and mevalonic acid lactone. Further, it is used to prepare 3-buten-2-one through dehydration over an anatase titanium dioxide catalyst. It is also employed in the preparation of 4-methyl-5-hydroxymethylthiazole, (±)-lineatin and beta-hydroxylactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-2-butanone is used in the preparation of fused benzazepine molecules as selective D3 receptor antagonists with pharmaceutical activity. It is also used in the preparation of verrucarin and mevalonic acid lactone. Further, it is used to prepare 3-buten-2-one through dehydration over an anatase titanium dioxide catalyst. It is also employed in the preparation of 4-methyl-5-hydroxymethylthiazole, (±)-lineatin and beta-hydroxylactones.

Solubility
Miscible with water, alcohol, ethanol and ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Hydroxy-2-butanone
1.02
89°C (192°F)
1.431
25 g
4-hydroxy-2-butanone, 2-butanone, 4-hydroxy, methylolacetone, 3-oxo-1-butanol, 3-oxobutanol, 3-ketobutan-1-ol, monomethylolacetone, 4-butanol-2-one, unii-tcm0bj44mf, 2-hydroxyethyl methyl ketone
LVSQXDHWDCMMRJ-UHFFFAOYSA-N
4-hydroxybutan-2-one
111509
88.11
590-90-9
73°C to 76°C (12 mmHg)
C4H8O2
MFCD00059005
1737666
Miscible with water,alcohol,ethanol and ether.
CC(=O)CCO
88.106
CHEBI:41268
95%

Chemical Identifiers

590-90-9
88.106
LVSQXDHWDCMMRJ-UHFFFAOYSA-N
111509
4-hydroxybutan-2-one
C4H8O2
MFCD00059005
4-hydroxy-2-butanone, 2-butanone, 4-hydroxy, methylolacetone, 3-oxo-1-butanol, 3-oxobutanol, 3-ketobutan-1-ol, monomethylolacetone, 4-butanol-2-one, unii-tcm0bj44mf, 2-hydroxyethyl methyl ketone
CHEBI:41268
CC(=O)CCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 209-693-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only