Learn More
4-Hydroxy-3,5-dimethoxyphenylacetic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2723103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Hydroxy-3,5-dimethoxyphenylacetic acid | |
127°C to 131°C | |
MFCD00016834 | |
2851658 | |
BQBQKSSTFGCRQL-UHFFFAOYSA-N | |
2-(4-hydroxy-3,5-dimethoxyphenyl)acetic acid | |
78093 | |
97% |
4385-56-2 | |
C10H12O5 | |
1 g | |
3,5-dimethoxy-4-hydroxyphenylacetic acid, homosyringic acid, 2-4-hydroxy-3,5-dimethoxyphenyl acetic acid, 4-hydroxy-3,5-dimethoxyphenyl acetic acid, 4-hydroxy-3,5-dimethoxyphenylacetic acid, benzeneacetic acid, 4-hydroxy-3,5-dimethoxy, acmc-1aewn, benzeneacetic acid,4-hydroxy-3,5-dimethoxy, 4-hydroxy-3,5-dimethoxy-phenyl-acetic acid, 4-hydroxy-3,5-dimethoxyphenyl acetic acid # | |
COC1=CC(=CC(=C1O)OC)CC(=O)O | |
212.201 | |
212.21 |
Chemical Identifiers
4385-56-2 | |
212.201 | |
BQBQKSSTFGCRQL-UHFFFAOYSA-N | |
78093 | |
COC1=CC(=CC(=C1O)OC)CC(=O)O |
C10H12O5 | |
MFCD00016834 | |
3,5-dimethoxy-4-hydroxyphenylacetic acid, homosyringic acid, 2-4-hydroxy-3,5-dimethoxyphenyl acetic acid, 4-hydroxy-3,5-dimethoxyphenyl acetic acid, 4-hydroxy-3,5-dimethoxyphenylacetic acid, benzeneacetic acid, 4-hydroxy-3,5-dimethoxy, acmc-1aewn, benzeneacetic acid,4-hydroxy-3,5-dimethoxy, 4-hydroxy-3,5-dimethoxy-phenyl-acetic acid, 4-hydroxy-3,5-dimethoxyphenyl acetic acid # | |
2-(4-hydroxy-3,5-dimethoxyphenyl)acetic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 224-494-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only