missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-4'-nitrobiphenyl 98.0+%, TCI America™
$132.35 - $1668.33
Chemical Identifiers
| CAS | 3916-44-7 |
|---|---|
| Molecular Formula | C12H9NO3 |
| Molecular Weight (g/mol) | 215.21 |
| MDL Number | MFCD00047742 |
| InChI Key | ZNDJDQOECGBUNK-UHFFFAOYSA-N |
| PubChem CID | 138078 |
| IUPAC Name | 4'-nitro-[1,1'-biphenyl]-4-ol |
| SMILES | OC1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O |
Chemical Identifiers
| 3916-44-7 | |
| 215.21 | |
| ZNDJDQOECGBUNK-UHFFFAOYSA-N | |
| 4'-nitro-[1,1'-biphenyl]-4-ol |
| C12H9NO3 | |
| MFCD00047742 | |
| 138078 | |
| OC1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O |
Specifications
| 3916-44-7 | |
| Yellow | |
| MFCD00047742 | |
| 4′C-Nitrobiphenyl-4-ol | |
| OC1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O | |
| 215.21 | |
| 215.21 | |
| Crystalline Powder |
| 207°C | |
| C12H9NO3 | |
| 1 g | |
| ZNDJDQOECGBUNK-UHFFFAOYSA-N | |
| 4'-nitro-[1,1'-biphenyl]-4-ol | |
| 138078 | |
| ≥98.0% (GC) | |
| 4-Hydroxy-4′-nitrobiphenyl |
Safety and Handling
TSCA : No