missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-6-methyl-nicotinic acid, 97%, Thermo Scientific Chemicals

$383.49 - $1508.42

Chemical Identifiers

CAS 33821-58-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD04038153
InChI Key AOJLDZLRTUWFFY-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methylnicotinic acid, 6-methyl-4-oxo-1,4-dihydropyridine-3-carboxylic acid, 4-hydroxy-6-methyl-nicotinic acid, 4-hydroxy 6-methylnicotinic acid, 4-hydroxy-6-methylpyridine-3-carboxylic acid, 4-hydroxy-6-methyl-3-pyridinecarboxylic acid, 6-methyl-4-hydroxynicotinic acid, chembrdg-bb 4002882, 6-methyl-4 1h-pyridone-3-carboxylic acid, 4-hydroxy-6-methyl-3-picolinic acid
PubChem CID 10702014
IUPAC Name 6-methyl-4-oxo-1H-pyridine-3-carboxylic acid
SMILES CC1=CC(=O)C(=CN1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6649903
View Documents
Thermo Scientific Chemicals
H6649903
1 g
Each for $383.49
 
AAH6649906
View Documents
Thermo Scientific Chemicals
H6649906
5 g
Each for $1,508.42
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33821-58-8
153.137
AOJLDZLRTUWFFY-UHFFFAOYSA-N
10702014
CC1=CC(=O)C(=CN1)C(=O)O
C7H7NO3
MFCD04038153
4-hydroxy-6-methylnicotinic acid, 6-methyl-4-oxo-1,4-dihydropyridine-3-carboxylic acid, 4-hydroxy-6-methyl-nicotinic acid, 4-hydroxy 6-methylnicotinic acid, 4-hydroxy-6-methylpyridine-3-carboxylic acid, 4-hydroxy-6-methyl-3-pyridinecarboxylic acid, 6-methyl-4-hydroxynicotinic acid, chembrdg-bb 4002882, 6-methyl-4 1h-pyridone-3-carboxylic acid, 4-hydroxy-6-methyl-3-picolinic acid
6-methyl-4-oxo-1H-pyridine-3-carboxylic acid

Specifications

33821-58-8
MFCD04038153
387402
AOJLDZLRTUWFFY-UHFFFAOYSA-N
6-methyl-4-oxo-1H-pyridine-3-carboxylic acid
10702014
97%
4-Hydroxy-6-methyl-nicotinic acid
C7H7NO3
1 g
4-hydroxy-6-methylnicotinic acid, 6-methyl-4-oxo-1,4-dihydropyridine-3-carboxylic acid, 4-hydroxy-6-methyl-nicotinic acid, 4-hydroxy 6-methylnicotinic acid, 4-hydroxy-6-methylpyridine-3-carboxylic acid, 4-hydroxy-6-methyl-3-pyridinecarboxylic acid, 6-methyl-4-hydroxynicotinic acid, chembrdg-bb 4002882, 6-methyl-4 1h-pyridone-3-carboxylic acid, 4-hydroxy-6-methyl-3-picolinic acid
CC1=CC(=O)C(=CN1)C(=O)O
153.137
153.14
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only