Learn More
4-Hydroxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
$96.79 - $294.96
Chemical Identifiers
CAS | 71597-85-8 |
---|---|
Molecular Formula | C6H7BO3 |
Molecular Weight (g/mol) | 137.93 |
MDL Number | MFCD01074628 |
InChI Key | COIQUVGFTILYGA-UHFFFAOYSA-N |
Synonym | 4-hydroxyphenyl boronic acid, 4-hydroxybenzeneboronic acid, 4-boronophenol, 4-hydroxyphenyl boranediol, p-hydroxyphenylboronic acid, 4-hydroxyphenyl-boronic acid, boronic acid, 4-hydroxyphenyl, pubchem1729, acmc-209ojb |
PubChem CID | 2734360 |
IUPAC Name | (4-hydroxyphenyl)boronic acid |
SMILES | OB(O)C1=CC=C(O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1559403
|
Thermo Scientific Chemicals
L1559403 |
1 g |
Each for $96.79
|
|
|||||
AAL1559406
|
Thermo Scientific Chemicals
L1559406 |
5 g |
Each for $294.96
|
|
|||||
Description
4-Hydroxybenzeneboronic acid is used for Suzuki-Miyaura coupling and Stille coupling and potential for introducing different alkyl groups, palladium-catalyzed aminocarbonylation and cross-coupling reactions, bio-supported palladium nanoparticles as phosphine-free catalyst for Suzuki reaction in water and Cu2O-catalyzed aerobic oxidative cross-coupling of tetrazoles. It is also used in preparation of rod-like dendronized polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxybenzeneboronic acid is used for Suzuki-Miyaura coupling and Stille coupling and potential for introducing different alkyl groups, palladium-catalyzed aminocarbonylation and cross-coupling reactions, bio-supported palladium nanoparticles as phosphine-free catalyst for Suzuki reaction in water and Cu2O-catalyzed aerobic oxidative cross-coupling of tetrazoles. It is also used in preparation of rod-like dendronized polymers.
Solubility
Soluble in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
71597-85-8 | |
137.93 | |
COIQUVGFTILYGA-UHFFFAOYSA-N | |
2734360 | |
OB(O)C1=CC=C(O)C=C1 |
C6H7BO3 | |
MFCD01074628 | |
4-hydroxyphenyl boronic acid, 4-hydroxybenzeneboronic acid, 4-boronophenol, 4-hydroxyphenyl boranediol, p-hydroxyphenylboronic acid, 4-hydroxyphenyl-boronic acid, boronic acid, 4-hydroxyphenyl, pubchem1729, acmc-209ojb | |
(4-hydroxyphenyl)boronic acid |
Specifications
71597-85-8 | |
C6H7BO3 | |
1 g | |
4-hydroxyphenyl boronic acid, 4-hydroxybenzeneboronic acid, 4-boronophenol, 4-hydroxyphenyl boranediol, p-hydroxyphenylboronic acid, 4-hydroxyphenyl-boronic acid, boronic acid, 4-hydroxyphenyl, pubchem1729, acmc-209ojb | |
COIQUVGFTILYGA-UHFFFAOYSA-N | |
(4-hydroxyphenyl)boronic acid | |
2734360 | |
97% |
∽285°C (decomposition) | |
MFCD01074628 | |
2963491 | |
Soluble in water | |
OB(O)C1=CC=C(O)C=C1 | |
137.93 | |
137.93 | |
4-Hydroxybenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only