Learn More
4-Hydroxybenzoic Acid, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 120991000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Hydroxybenzoic acid | |
99-96-7 | |
100.0 | |
Ivory to White | |
Authentic | |
Plastic bottle | |
HOC6H4CO2H | |
100 g | |
15, 4850 | |
Solubility in water: 5g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in chloroform,practically insoluble in carbon disulfide | |
OC(=O)C1=CC=C(O)C=C1 | |
138.12 | |
CHEBI:30763 | |
99+% |
99% | |
99.0 | |
214°C to 217°C | |
199°C | |
99% min. (HPLC) | |
C7H6O3 | |
MFCD00002547 | |
10, 149 | |
p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german | |
FJKROLUGYXJWQN-UHFFFAOYSA-N | |
4-hydroxybenzoic acid | |
135 | |
138.12 | |
Crystalline Powder |
Chemical Identifiers
99-96-7 | |
138.12 | |
FJKROLUGYXJWQN-UHFFFAOYSA-N | |
135 | |
4-hydroxybenzoic acid |
C7H6O3 | |
MFCD00002547 | |
p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german | |
CHEBI:30763 | |
OC(=O)C1=CC=C(O)C=C1 |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 202-804-9
RTECSNumber : DH1925000
TSCA : TSCA
RUO – Research Use Only