missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxybenzoic Acid, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 120995000

 View more versions of this product

Catalog No. AC120995000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Hydroxybenzoic acid
99-96-7
100.0
Ivory to White
Authentic
Plastic bottle
HOC6H4CO2H
500 g
15, 4850
Solubility in water: 5g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in chloroform,practically insoluble in carbon disulfide
OC(=O)C1=CC=C(O)C=C1
138.12
CHEBI:30763
99+%
99%
99.0
214.0°C to 217.0°C
199°C
99% min. (HPLC)
C7H6O3
MFCD00002547
10, 149
p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german
FJKROLUGYXJWQN-UHFFFAOYSA-N
4-hydroxybenzoic acid
135
138.12
Crystalline Powder

Chemical Identifiers

99-96-7
138.12
FJKROLUGYXJWQN-UHFFFAOYSA-N
135
4-hydroxybenzoic acid
C7H6O3
MFCD00002547
p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german
CHEBI:30763
OC(=O)C1=CC=C(O)C=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 202-804-9

RTECSNumber : DH1925000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only