missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxybenzoic Acid Hydrazide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 121000250

 View more versions of this product

Catalog No. AC121000250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Hydroxybenzoic acid hydrazide
97.5
260.0°C to 264.0°C
Authentic
Plastic bottle
HOC6H4CONHNH2
25 g
4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide
ZMZGIVVRBMFZSG-UHFFFAOYSA-N
4-hydroxybenzohydrazide
1742
98%
5351-23-5
100.0
Beige to White
98%
C7H8N2O2
MFCD00007605
10, 174
Solubility in water: 26.5g/L (25°C). Other solubilities: soluble in methanol,ethanol
C1=CC(=CC=C1C(=O)NN)O
152.15
152.15
Fine Crystalline Powder

Chemical Identifiers

5351-23-5
152.15
ZMZGIVVRBMFZSG-UHFFFAOYSA-N
1742
C1=CC(=CC=C1C(=O)NN)O
C7H8N2O2
MFCD00007605
4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide
4-hydroxybenzohydrazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye pr

GHS Signal Word: Warning

EINECSNumber : 226-326-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only