Learn More
4-Hydroxydiphenylamine, 98%, Thermo Scientific Chemicals
$77.10 - $271.47
Chemical Identifiers
CAS | 122-37-2 |
---|---|
Molecular Formula | C12H11NO |
Molecular Weight (g/mol) | 185.226 |
MDL Number | MFCD00020142 |
InChI Key | JTTMYKSFKOOQLP-UHFFFAOYSA-N |
Synonym | 4-hydroxydiphenylamine, 4-phenylamino phenol, p-anilinophenol, phenol, 4-phenylamino, p-hydroxydiphenylamine, p-oxydiphenylamine, phenyl-p-aminophenol, n-phenyl-p-aminophenol, 4-phenylaminophenol, p-hydroxydifenylamin |
PubChem CID | 31208 |
IUPAC Name | 4-anilinophenol |
SMILES | C1=CC=C(C=C1)NC2=CC=C(C=C2)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1894509
|
Thermo Scientific Chemicals
L1894509 |
10 g |
N/A
|
|
|||||
AAL1894518
|
Thermo Scientific Chemicals
L1894518 |
50 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
122-37-2 | |
185.226 | |
JTTMYKSFKOOQLP-UHFFFAOYSA-N | |
31208 | |
C1=CC=C(C=C1)NC2=CC=C(C=C2)O |
C12H11NO | |
MFCD00020142 | |
4-hydroxydiphenylamine, 4-phenylamino phenol, p-anilinophenol, phenol, 4-phenylamino, p-hydroxydiphenylamine, p-oxydiphenylamine, phenyl-p-aminophenol, n-phenyl-p-aminophenol, 4-phenylaminophenol, p-hydroxydifenylamin | |
4-anilinophenol |
Specifications
122-37-2 | |
215°C to 218°C (12 mmHg) | |
Odorless | |
MFCD00020142 | |
511942 | |
JTTMYKSFKOOQLP-UHFFFAOYSA-N | |
4-anilinophenol | |
31208 | |
98% |
66°C to 71°C | |
209°C (408°F) | |
C12H11NO | |
10 g | |
4-hydroxydiphenylamine, 4-phenylamino phenol, p-anilinophenol, phenol, 4-phenylamino, p-hydroxydiphenylamine, p-oxydiphenylamine, phenyl-p-aminophenol, n-phenyl-p-aminophenol, 4-phenylaminophenol, p-hydroxydifenylamin | |
C1=CC=C(C=C1)NC2=CC=C(C=C2)O | |
185.226 | |
185.23 | |
4-Hydroxydiphenylamine |
Safety and Handling
GHS H Statement
H311-H318-H315-H335
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
EINECSNumber : 204-538-9
RTECSNumber : SJ6950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only