missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Hydroxymethyl)benzoic acid, 98+%, Thermo Scientific Chemicals
$115.79 - $1406.15
Chemical Identifiers
CAS | 3006-96-0 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD00017598 |
InChI Key | WWYFPDXEIFBNKE-UHFFFAOYSA-N |
Synonym | 4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd |
PubChem CID | 76360 |
IUPAC Name | 4-(hydroxymethyl)benzoic acid |
SMILES | OCC1=CC=C(C=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2218303
|
Thermo Scientific Chemicals
B2218303 |
1 g |
Each for $115.79
|
|
||||||
AAB2218306
|
Thermo Scientific Chemicals
B2218306 |
5 g |
Each for $359.75
|
|
||||||
AAB2218314
|
Thermo Scientific Chemicals
B2218314 |
25 g |
Each for $1,406.15
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3006-96-0 | |
152.15 | |
WWYFPDXEIFBNKE-UHFFFAOYSA-N | |
76360 | |
OCC1=CC=C(C=C1)C(O)=O |
C8H8O3 | |
MFCD00017598 | |
4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd | |
4-(hydroxymethyl)benzoic acid |
Specifications
3006-96-0 | |
Odorless | |
MFCD00017598 | |
2690023 | |
WWYFPDXEIFBNKE-UHFFFAOYSA-N | |
4-(hydroxymethyl)benzoic acid | |
76360 | |
≥98% |
179°C to 183°C | |
C8H8O3 | |
1 g | |
4-hydroxymethyl benzoic acid, 4-hydroxythylbenzoic acid, unii-9z28l8sthd, hmba linker, 4-hydroxymethyl-benzoic acid, benzoic acid, 4-hydroxymethyl, p-hydroxymethyl benzoic acid, 4-hydroxymethyl benzoicacid, 9z28l8sthd | |
OCC1=CC=C(C=C1)C(O)=O | |
152.15 | |
152.15 | |
4-(Hydroxymethyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only