missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxyphenylacetamide, 99%, Thermo Scientific Chemicals

$54.25 - $159.05

Chemical Identifiers

CAS 17194-82-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00017145
InChI Key YBPAYPRLUDCSEY-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
PubChem CID 86986
IUPAC Name 2-(4-hydroxyphenyl)acetamide
SMILES C1=CC(=CC=C1CC(=O)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1526609
View Documents
Thermo Scientific Chemicals
A1526609
10 g
Each for $54.25
 
AAA1526618
View Documents
Thermo Scientific Chemicals
A1526618
50 g
Each for $159.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17194-82-0
151.165
YBPAYPRLUDCSEY-UHFFFAOYSA-N
86986
C1=CC(=CC=C1CC(=O)N)O
C8H9NO2
MFCD00017145
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
2-(4-hydroxyphenyl)acetamide

Specifications

17194-82-0
C8H9NO2
10 g
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4
C1=CC(=CC=C1CC(=O)N)O
151.165
151.17
4-Hydroxyphenylacetamide
173°C to 175°C
MFCD00017145
2085984
YBPAYPRLUDCSEY-UHFFFAOYSA-N
2-(4-hydroxyphenyl)acetamide
86986
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 241-235-0

RTECSNumber : AC3851500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only