missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hydroxyphenylacetamide 98.0+%, TCI America™
Supplier: TCI America H0853500G
Specifications
4-Hydroxyphenylacetamide | |
177°C | |
MFCD00017145 | |
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4 | |
C1=CC(=CC=C1CC(=O)N)O | |
151.165 | |
151.17 | |
Crystalline Powder |
17194-82-0 | |
C8H9NO2 | |
500 g | |
YBPAYPRLUDCSEY-UHFFFAOYSA-N | |
2-(4-hydroxyphenyl)acetamide | |
86986 | |
≥98.0% (HPLC,N) |
Chemical Identifiers
17194-82-0 | |
151.165 | |
YBPAYPRLUDCSEY-UHFFFAOYSA-N | |
86986 | |
C1=CC(=CC=C1CC(=O)N)O |
C8H9NO2 | |
MFCD00017145 | |
4-hydroxyphenylacetamide, 2-4-hydroxyphenyl acetamide, p-hydroxyphenylacetamide, 4-hydroxybenzeneacetamide, benzeneacetamide, 4-hydroxy, p-carbamoylmethyl phenol, p-hydroxyphenyl acetamide, 2-p-hydroxyphenyl acetamide, 4-hydroxyphenyl acetamide, unii-0hy0n4itn4 | |
2-(4-hydroxyphenyl)acetamide |
Safety and Handling
EINECSNumber : (3)-3726
RTECSNumber : AC3851500
TSCA : Yes