missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodo-2-nitrotoluene, 97%, Thermo Scientific Chemicals

$199.05 - $737.78

Chemical Identifiers

CAS 41252-97-5
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00051090
InChI Key QLMRDNPXYNJQMQ-UHFFFAOYSA-N
Synonym 4-iodo-2-nitrotoluene, benzene, 4-iodo-1-methyl-2-nitro, pubchem15938, 2-nitro-4-iodotoluene, acmc-1apfk, maybridge1_008109, ksc493s9d, 4-iodo-1-methyl-2-nitro-benzene
PubChem CID 170481
IUPAC Name 4-iodo-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)I)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1860706
View Documents
Thermo Scientific Chemicals
A1860706
5 g
Each for $199.05
 
AAA1860714
View Documents
Thermo Scientific Chemicals
A1860714
25 g
Each for $737.78
 
Description

Description

4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications

Chemical Identifiers

41252-97-5
263.034
QLMRDNPXYNJQMQ-UHFFFAOYSA-N
170481
CC1=C(C=C(C=C1)I)[N+](=O)[O-]
C7H6INO2
MFCD00051090
4-iodo-2-nitrotoluene, benzene, 4-iodo-1-methyl-2-nitro, pubchem15938, 2-nitro-4-iodotoluene, acmc-1apfk, maybridge1_008109, ksc493s9d, 4-iodo-1-methyl-2-nitro-benzene
4-iodo-1-methyl-2-nitrobenzene

Specifications

41252-97-5
C7H6INO2
5 g
4-iodo-2-nitrotoluene, benzene, 4-iodo-1-methyl-2-nitro, pubchem15938, 2-nitro-4-iodotoluene, acmc-1apfk, maybridge1_008109, ksc493s9d, 4-iodo-1-methyl-2-nitro-benzene
QLMRDNPXYNJQMQ-UHFFFAOYSA-N
4-iodo-1-methyl-2-nitrobenzene
170481
97%
58°C to 62°C
MFCD00051090
Light sensitive
Slightly soluble in water.
CC1=C(C=C(C=C1)I)[N+](=O)[O-]
263.034
263.03
4-Iodo-2-nitrotoluene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 255-279-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only