missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodo-3-methyl-5-phenylisoxazole, 97%, Thermo Scientific™

$47.08 - $47.08

Chemical Identifiers

CAS 16114-53-7
Molecular Formula C10H8INO
Molecular Weight (g/mol) 285.084
MDL Number MFCD03086126
InChI Key RLZGZJOMCXKYIW-UHFFFAOYSA-N
Synonym 4-iodo-3-methyl-5-phenylisoxazole, isoxazole,4-iodo-3-methyl-5-phenyl, 3-methyl-5-phenyl-4-iodoisoxazole, 4-iodo-3-methylisoxazol-5-yl benzene, 4-iodo-3-methyl-1,2-oxazol-5-yl benzene
PubChem CID 2776520
IUPAC Name 4-iodo-3-methyl-5-phenyl-1,2-oxazole
SMILES CC1=NOC(=C1I)C2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC20311DA
SDS View Documents
Thermo Scientific Chemicals
CC20311DA
1 g Amber Glass Bottle
Each for $47.08
 
Description

Description

Specifications

Chemical Identifiers

16114-53-7
285.084
RLZGZJOMCXKYIW-UHFFFAOYSA-N
2776520
CC1=NOC(=C1I)C2=CC=CC=C2
C10H8INO
MFCD03086126
4-iodo-3-methyl-5-phenylisoxazole, isoxazole,4-iodo-3-methyl-5-phenyl, 3-methyl-5-phenyl-4-iodoisoxazole, 4-iodo-3-methylisoxazol-5-yl benzene, 4-iodo-3-methyl-1,2-oxazol-5-yl benzene
4-iodo-3-methyl-5-phenyl-1,2-oxazole

Specifications

16114-53-7
100.0
Amber Glass Bottle
MFCD03086126
4-iodo-3-methyl-5-phenylisoxazole, isoxazole,4-iodo-3-methyl-5-phenyl, 3-methyl-5-phenyl-4-iodoisoxazole, 4-iodo-3-methylisoxazol-5-yl benzene, 4-iodo-3-methyl-1,2-oxazol-5-yl benzene
CC1=NOC(=C1I)C2=CC=CC=C2
285.084
285.08
4-Iodo-3-methyl-5-phenylisoxazole
96.5
97%
C10H8INO
1 g
RLZGZJOMCXKYIW-UHFFFAOYSA-N
4-iodo-3-methyl-5-phenyl-1,2-oxazole
2776520
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'